missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Aspartate Transaminase Polyclonal antibody specifically detects Aspartate Transaminase in Porcine samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Aspartate Transaminase |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | aspartate aminotransferase, cytoplasmic, CASPAT |
| Host Species | Sheep |
| Immunogen | Aspartate Transaminase Antibody was produced by repeated immunizations with Aspartate Transaminase from pig heart. (Uniprot: P00503) |
| Purification Method | Multi-step |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?