missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aspartate Transaminase Antibody [CoraFluor™ 1], Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-78089CL1
This item is not returnable.
View return policy
Description
Aspartate Transaminase Polyclonal antibody specifically detects Aspartate Transaminase in Porcine samples. It is validated for ELISA,Western Blot
Specifications
| Aspartate Transaminase | |
| Polyclonal | |
| PBS | |
| Sheep | |
| Multi-step | |
| RUO | |
| 396967 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| aspartate aminotransferase, cytoplasmic, CASPAT | |
| Aspartate Transaminase Antibody was produced by repeated immunizations with Aspartate Transaminase from pig heart. (Uniprot: P00503) | |
| 0.1 mL | |
| Primary | |
| Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction