missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aspartate Aminotransferase Rabbit anti-Human, Mouse, Rat, Clone: 5J1B4, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £413.00
Specifications
| Antigen | Aspartate Aminotransferase |
|---|---|
| Clone | 5J1B4 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18372066
|
Novus Biologicals
NBP3-15347-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18327604
|
Novus Biologicals
NBP3-15347-100UL |
100 μg |
£413.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aspartate Aminotransferase Monoclonal antibody specifically detects Aspartate Aminotransferase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| Aspartate Aminotransferase | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| aspartate aminotransferase, cytoplasmic, ASTQTL1, CASPAT, CCAT, Cysteine aminotransferase, cytoplasmic, Cysteine transaminase, cytoplasmic, EC 2.6.1.1, EC 2.6.1.3, GIG18, Glutamate oxaloacetate transaminase 1, glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1), GOT1, growth-inhibiting protein 18, Transaminase A | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human Aspartate Aminotransferase (P17174). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 5J1B4 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2805 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title