missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aspartate Aminotransferase Antibody (3G11), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59483PCP
This item is not returnable.
View return policy
Description
Aspartate Aminotransferase Monoclonal antibody specifically detects Aspartate Aminotransferase in Human samples. It is validated for Flow Cytometry
Specifications
| Aspartate Aminotransferase | |
| Monoclonal | |
| PerCP | |
| aspartate aminotransferase, cytoplasmic, ASTQTL1, CASPAT, CCAT, Cysteine aminotransferase, cytoplasmic, Cysteine transaminase, cytoplasmic, EC 2.6.1.1, EC 2.6.1.3, GIG18, Glutamate oxaloacetate transaminase 1, glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1), GOT1, growth-inhibiting protein 18, Transaminase A | |
| Recombinant human Aspartate Aminotransferase (1-413aa) purified from E. coli | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 2805 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| 3G11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction