missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Asparagine Synthetase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102802
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Asparagine synthetase.
ASNS (asparagine synthetase [glutamine-hydrolyzing]) is involved in the subpathway that synthesizes f L-asparagine from L-aspartate. Mutations affecting the gene can result in asparagine synthetase deficiency (ASNSD).
Specifications
| Asparagine Synthetase | |
| Polyclonal | |
| Unconjugated | |
| Asns | |
| ASNS; ASNSD; asparagine synthetase; asparagine synthetase (glutamine-hydrolyzing); asparagine synthetase [glutamine-hydrolyzing]; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase; TS11; TS11 cell cycle control protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25612, 27053, 440 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P08243, P49088, Q61024 | |
| Asns | |
| A synthesized peptide derived from human ASNS(Accession P08243), corresponding to amino acid residues F8-G58. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction