missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asparagine synthetase Antibody (OTI1A10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70215
This item is not returnable.
View return policy
Description
Asparagine synthetase Monoclonal specifically detects Asparagine synthetase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Asparagine synthetase | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| asparagine synthetase, asparagine synthetase (glutamine-hydrolyzing), asparagine synthetase [glutamine-hydrolyzing], Cell cycle control protein TS11, EC 6.3.5.4, Glutamine-dependent asparagine synthetase, TS11, TS11 cell cycle control protein | |
| Mouse | |
| 64.2 kDa | |
| 100 μg | |
| 440 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1A10 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| ASNS | |
| Full length human recombinant protein of human ASNS(NP_597680) produced in HEK293T cell. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction