missing translation for 'onlineSavingsMsg'
Learn More

Asparagine synthetase Antibody (3B3), Novus Biologicals™

Product Code. 18375047 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375047 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375047 Supplier Novus Biologicals Supplier No. H00000440M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Asparagine synthetase Monoclonal antibody specifically detects Asparagine synthetase in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Asparagine synthetase
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3B3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH14621
Gene Alias asparagine synthetase, asparagine synthetase (glutamine-hydrolyzing), asparagine synthetase [glutamine-hydrolyzing], Cell cycle control protein TS11, EC 6.3.5.4, Glutamine-dependent asparagine synthetase, TS11, TS11 cell cycle control protein
Host Species Mouse
Immunogen ASNS (AAH14621, 281 a.a. ~ 380 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YPLQTFAIGMEDSPDLLAARKVADHIGSEHYEVLFNSEEGIQALDEVIFSLETYDITTVRASVGMYLISKYIRKNTDSVVIFSGEGSDELTQGYIYFHKA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 440
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.