missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13244-1-AP-20UL
This item is not returnable.
View return policy
Description
ASPA is an enzyme that catalyzes the conversion of N-acetyl_L-aspartic acid (NAA) to aspartate and acetate. NAA is abundant in the brain where hydrolysis by aspartoacylase is thought to help maintain white matter. This protein is an NAA scavenger in other tissues.Specifications
| ASPA | |
| Polyclonal | |
| Unconjugated | |
| ASPA | |
| ACY 2, ACY2, Aminoacylase 2, ASP, ASPA, Aspartoacylase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11484, 443, 79251 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P45381, Q8R3P0, Q9R1T5 | |
| ASPA | |
| ASPA Fusion Protein Ag4029 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction