missing translation for 'onlineSavingsMsg'
Learn More

ASP3026, MedChemExpress

Product Code. 16446166
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446166 100 mg 100mg
16441817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16446166 Supplier Medchem Express Supplier No. HY13326

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ASP3026 is a potent, selective and orally active inhibitor of anaplastic lymphoma kinase (ALK). ASP3026 induces apoptosis of tumor cells. ASP3026 can be used for the research of non-small cell lung cancer (NSCLC).

ASP3026 is a potent, selective and orally active inhibitor of anaplastic lymphoma kinase (ALK). ASP3026 induces apoptosis of tumor cells. ASP3026 can be used for the research of non-small cell lung cancer (NSCLC).

Specifications

Chemical Name or Material ASP3026
CAS 1097917-15-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H40N8O3S
Quantity 100 mg
Solubility Information DMSO : 20 mg/mL (34.44 mM; Need ultrasonic)
SMILES COC(C=C(N1CCC(N2CCN(C)CC2)CC1)C=C3)=C3NC4=NC=NC(NC5=C(S(C(C)C)(=O)=O)C=CC=C5)=N4
Molecular Weight (g/mol) 580.74
Formula Weight 580.74
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.