missing translation for 'onlineSavingsMsg'
Learn More

Asp-AMS, MedChemExpress

Product Code. 16437437
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16437437

Brand: Medchem Express HY112860

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Asp-AMS, an analogue of aspartyl-adenylate, is an aspartyl-tRNA synthetase inhibitor and also a strong competitive inhibitor of the mitochondrial enzyme.

Asp-AMS, an analogue of aspartyl-adenylate, is an aspartyl-tRNA synthetase inhibitor and also a strong competitive inhibitor of the mitochondrial enzyme.

Specifications

Chemical Name or Material Asp-AMS
CAS 828288-98-8
Color White
Purity Grade Notes Research
Molecular Formula C14H19N7O9S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (216.73 mM)
SMILES O[C@@H]([C@H]([C@H](N1C=NC2=C1N=CN=C2N)O3)O)[C@H]3COS(=O)(NC([C@@H](N)CC(O)=O)=O)=O
Molecular Weight (g/mol) 461.41
Formula Weight 461.41
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.