missing translation for 'onlineSavingsMsg'
Learn More

ASP-9521, MedChemExpress

Product Code. 16480418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480418 5 mg 5mg
16467746 100 mg 100mg
16463887 50 mg 50mg
16461296 200 mg 200mg
16447958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16480418 Supplier Medchem Express Supplier No. HY19903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ASP-9521 is a potent, selective and orally available AKR1C3 inhibitor with an IC50 of 11 nM for human AKR1C3.

ASP-9521 is a potent, selective and orally available AKR1C3 inhibitor with an IC50 of 11 nM for human AKR1C3.

Specifications

Chemical Name or Material ASP-9521
CAS 1126084-37-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H26N2O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (302.65 mM; Need ultrasonic)
SMILES O=C(N1CCC(CC(C)(O)C)CC1)C(N2)=CC3=C2C=CC(OC)=C3
Molecular Weight (g/mol) 330.42
Formula Weight 330.42
Percent Purity 96.39%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.