missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ASP 3026
GREENER_CHOICE

Product Code. p-7110809
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18701698

Brand: Tocris Bioscience™ 5310/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent anaplastic lymphoma kinase (ALK) inhibitor; also potent ACK inhibitor

ASP 3026 is a potent anaplastic lymphoma kinase (ALK) inhibitor (IC50 = 3.5 nM). Also inhibits Ack and ROS1 activity (IC50 values are 5.8 and 8.9 nM respectively). Attenuates proliferation, and induces apoptosis of NPM-ALK+ T cell anaplastic large-cell lymphoma (ALCL) cells in vitro. Suppresses tumor growth of NPM-ALK+ ALCL cell xenografts in mice. Causes tumor shrinkage in hEML4-ALK transgenic mice. Orally available.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N2-[2-Methoxy-4-[4-(4-methyl-1-piperazinyl)-1-piperidinyl]phenyl]-N4-[2-[(1-methylethyl)sulfonyl]phenyl]-1,3,5-triazine-2,4-diamine
CAS 1097917-15-1
Quantity 10 mg
Target ALK Inhibitors
Molecular Formula C29H40N8O3S
Formula Weight 580.74
Purity >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.