missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ASP 3026

Description
ASP 3026 is a potent anaplastic lymphoma kinase (ALK) inhibitor (IC50 = 3.5 nM). Also inhibits Ack and ROS1 activity (IC50 values are 5.8 and 8.9 nM respectively). Attenuates proliferation, and induces apoptosis of NPM-ALK+ T cell anaplastic large-cell lymphoma (ALCL) cells in vitro. Suppresses tumor growth of NPM-ALK+ ALCL cell xenografts in mice. Causes tumor shrinkage in hEML4-ALK transgenic mice. Orally available.
Specifications
Specifications
| Chemical Name or Material | N2-[2-Methoxy-4-[4-(4-methyl-1-piperazinyl)-1-piperidinyl]phenyl]-N4-[2-[(1-methylethyl)sulfonyl]phenyl]-1,3,5-triazine-2,4-diamine |
| CAS | 1097917-15-1 |
| Quantity | 50 mg |
| Target | ALK Inhibitors |
| Molecular Formula | C29H40N8O3S |
| Formula Weight | 580.74 |
| Purity | >99% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?