missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ASP 3026
GREENER_CHOICE

Product Code. 18701698
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18701698 10 mg 10mg
18730518 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18701698 Supplier Tocris Bioscience™ Supplier No. 5310/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent anaplastic lymphoma kinase (ALK) inhibitor; also potent ACK inhibitor

ASP 3026 is a potent anaplastic lymphoma kinase (ALK) inhibitor (IC50 = 3.5 nM). Also inhibits Ack and ROS1 activity (IC50 values are 5.8 and 8.9 nM respectively). Attenuates proliferation, and induces apoptosis of NPM-ALK+ T cell anaplastic large-cell lymphoma (ALCL) cells in vitro. Suppresses tumor growth of NPM-ALK+ ALCL cell xenografts in mice. Causes tumor shrinkage in hEML4-ALK transgenic mice. Orally available.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N2-[2-Methoxy-4-[4-(4-methyl-1-piperazinyl)-1-piperidinyl]phenyl]-N4-[2-[(1-methylethyl)sulfonyl]phenyl]-1,3,5-triazine-2,4-diamine
CAS 1097917-15-1
Quantity 10 mg
Target ALK Inhibitors
Molecular Formula C29H40N8O3S
Formula Weight 580.74
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.