missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASNSD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567372
This item is not returnable.
View return policy
Description
Immunogen sequence: KLRRTRICHLI RPLDTVLDDS IGCAVWFASR GIGWLVAQEG VKSYQSNAKV VLTGIGADEQ LAGYSRHRVR FQSHGLEGLN KE Highest antigen sequence identity to the following orthologs - mouse 84%, rat 82%.
ASNSD1 (Asparagine Synthetase Domain Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include asparagine synthase (glutamine-hydrolyzing) activity. An important paralog of this gene is ENSG00000286165. [GeneCards].
Specifications
| ASNSD1 | |
| Polyclonal | |
| Unconjugated | |
| ASNSD1 | |
| ASNSD1; asparagine synthetase domain containing 1; asparagine synthetase domain-containing protein 1; HCV NS3-transactivated protein 1; NBLA00058; NS3TP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54529 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NWL6 | |
| ASNSD1 | |
| Recombinant Human ASNSD1. Recombinant protein control fragment (Product #RP-108032). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction