missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ASNSD1 Polyclonal specifically detects ASNSD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | ASNSD1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q9NWL6 |
| Gene Alias | asparagine synthetase domain containing 1, asparagine synthetase domain-containing protein 1, HCV NS3-transactivated protein 1, NBLA00058, NS3TP1FLJ20752 |
| Gene Symbols | ASNSD1 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?