missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASNA1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518922
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
ASNA1 is the human homolog of the bacterial arsA gene. In E. coli, ArsA ATPase is the catalytic component of a multisubunit oxyanion pump that is responsible for resistance to arsenicals and antimonials.
Specifications
| ASNA1 | |
| Polyclonal | |
| Unconjugated | |
| GET3 | |
| 1810048H22Rik; ARSA; arsA arsenite transporter, ATP-binding, homolog 1; arsA arsenite transporter, ATP-binding, homolog 1 (bacterial); ARSA1; ARSA-I; Arsenical pump-driving ATPase; Arsenical resistance ATPase; arsenite-stimulated ATPase; arsenite-translocating ATPase; Arsenite-transporting ATPase; asna1; ASNA-I; ATPase asna1; ATPase GET3; GET3; guided entry of tail-anchored proteins factor 3, ATPase; hARSA-I; hASNA-I; MGC3821; si:dkey-121b10.2; Transmembrane domain recognition complex 40 kDa ATPase subunit; TRC40; wu:fd05d03; zgc:56540; zgc:56540 protein; zgc:86799 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 288919, 439, 56495 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O43681, O54984 | |
| GET3 | |
| Synthetic peptide sequence (PHEVRGADKVNT) corresponding to the internal amino acids of ASNA1 (aa 322-333). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction