missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASNA1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1B7C11, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ASNA1 |
|---|---|
| Clone | 1B7C11 |
| Concentration | 1 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16868135
|
Proteintech
66346-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16858135
|
Proteintech
66346-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASNA1 is the human homolog of the bacterial arsA gene. In E. coli, ArsA ATPase is the catalytic component of a multisubunit oxyanion pump that is responsible for resistance to arsenicals and antimonials.Specifications
| ASNA1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ARSA, ARSA I, ARSA1, Arsenical pump driving ATPase, Arsenite stimulated ATPase, ASNA1, ATPase ASNA1, hARSA I, hASNA I, TRC40 | |
| Asna1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1B7C11 | |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Pig, Rat, Mouse, Human | |
| O43681, O54984 | |
| 100520362, 288919, 439, 56495 | |
| ASNA1 Fusion Protein Ag7936 | |
| Primary | |
| -20°C | |
| ASNA1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title