missing translation for 'onlineSavingsMsg'
Learn More

ASNA1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP1-20889

Product Code. 18118059

  • £407.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ASNA1 Polyclonal specifically detects ASNA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ASNA1
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
ARSA, arsA arsenite transporter, ATP-binding, homolog 1 (bacterial), ARSA1, ARSA-I, Arsenical pump-driving ATPase, Arsenite-stimulated ATPase, ATPase ASNA1, EC 3.6, EC 3.6.3.16, GET3, golgi to ER traffic 3 homolog, hARSA-I, hASNA-I, MGC3821, Transmembrane domain recognition complex 40 kDa ATPase subunit, transmembrane domain recognition complex, 40kDa, TRC40arsA (bacterial) arsenite transporter, ATP-binding, homolog 1
Goat
Affinity Purified
RUO
Primary
Cross reacts with Human and mouse. Expected from sequence similarity: Rat, Dog, Pig and Cow.
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, ELISA
0.5 mg/mL
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:2000
NP_004308.2
ASNA1
Peptide with sequence C-PHEVRGADKVNT corresponding to internal region according to NP_004308.2.
0.1 mg
metabolism
439
Human, Mouse, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only