missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASMTL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554530
This item is not returnable.
View return policy
Description
Immunogen sequence: RLLDICAAMG LLEKTEQGYS NTETANVYLA SDGEYSLHGF IMHNNDLTWN LFTYLEFAIR EGTNQHHRAL GKKAEDLFQD AYYQSPETRL RFMRAMHGMT KLTACQVATA FNLSRFSSAC DVGGCTGALA RELAREYPRM QVTV Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 40%.
The protein encoded by this gene has an N-terminus that is similar to the multicopy associated filamentation (maf) protein of Bacillus subtilis and to orfE of Escherichia coli, while the C-terminus is similar to N-acetylserotonin O-methyltransferase. This gene is located in the pseudoautosomal region 1 (PAR1) of X and Y chromosomes. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| ASMTL | |
| Polyclonal | |
| Unconjugated | |
| ASMTL | |
| acetylserotonin N-methyltransferase-like; acetylserotonin O-methyltransferase-like; ASMTL; ASMTLX; ASMTLY; ASTML; dTTP/UTP pyrophosphatase; dTTPase/UTPase; N-acetylserotonin O-methyltransferase-like protein; Nucleoside triphosphate pyrophosphatase; Nucleotide PPase; Nucleotide pyrophosphatase; Probable bifunctional dTTP/UTP pyrophosphatase/methyltransferase protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8623 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95671 | |
| ASMTL | |
| Recombinant protein corresponding to Human ASMTL. Recombinant protein control fragment (Product #RP-92232). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction