missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASM Recombinant Rabbit Monoclonal Antibody (5T2N2)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558120

 View more versions of this product

Product Code. 30282694

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PARLHRIVPR LRDVFGWGNL TCPICKGLFT AINLGLKKEP NVARVGSVAI KLCNLLKIAP PAVCQSIVHL FEDDMVEVWR RSVLSPSEAC GLLLGSTCGH WDIFSSWNIS LPTVPKPPPK PPSPPAPGAP VSRILFLTDL HWDHDYLEGT DPDCADPLCC RRGSGLPPAS RPGAGYWGEY SKCDLPLRTL ESLLSGLGPA GPFDMVYWTG DIPAHDVWHQ TRQDQLRALT TVTALVRKFL GPVPVYPAVG NHESTPVNSF PPPFIEGNHS S.

The protein encoded by this gene is a lysosomal acid sphingomyelinase that converts sphingomyelin to ceramide. The encoded protein also has phospholipase C activity. Defects in this gene are a cause of Niemann-Pick disease type A (NPA) and Niemann-Pick disease type B (NPB). Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ASM
Recombinant Monoclonal
2.03 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P17405, Q04519
Smpd1
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin)
5T2N2
Unconjugated
Smpd1
Acid sphingomyelinase; ASM; aSMase; A-SMase; NPD; Smpd1; Sphingomyelin phosphodiesterase; sphingomyelin phosphodiesterase 1; sphingomyelin phosphodiesterase 1, acid lysosomal; Zn-SMase
Rabbit
Protein A
RUO
20597, 308909, 6609
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.