missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASL Monoclonal Antibody (OTI14C7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526708
This item is not returnable.
View return policy
Description
ASL Monoclonal Antibody for Western Blot
ASL encodes a member of the lyase 1 family. The encoded protein forms a cytosolic homotetramer and primarily catalyzes the reversible hydrolytic cleavage of argininosuccinate into arginine and fumarate, an essential step in the liver in detoxifying ammonia via the urea cycle. Mutations in this gene result in the autosomal recessive disorder argininosuccinic aciduria, or argininosuccinic acid lyase deficiency. A nontranscribed pseudogene is also located on the long arm of chromosome 22. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| ASL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04424 | |
| ASL | |
| Full length human recombinant protein of ASL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI14C7 | |
| Unconjugated | |
| ASL | |
| 2510006M18Rik; argininosuccinase; argininosuccinate lyase; Arginosuccinase; ASAL; Asl | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 435 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction