missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASK1 Monoclonal Antibody (K52022_9A7), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | ASK1 |
|---|---|
| Clone | K52022_9A7 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30193598
|
Invitrogen™
MA556141 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30205504
|
Invitrogen™
MA555761 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: DLDVEDDHEE QPSNQTVRRP QAVIEDAVAT SGVSTLSSTV SHDSQSAHRS LNVQLGRMKI ETNRLLEELV RKEKELQALL HRAIEEKDQE IKHLKLKSQP IEIPELPVFH
ASK1 Monoclonal antibody specifically detects ASK1 in Human samples. It is validated for Western BlotSpecifications
| ASK1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 7420452D20Rik; apoptosis signal regulating kinase 1; apoptosis signal-regulating kinase 1; ASK; ASK 1; ASK1; ASK-1; EC 2.7.11.25; kinase ASK1; M3K5; MAP/ERK kinase kinase 5; Map3k5; MAPK/ERK kinase kinase 5; MAPKKK5; MEK kinase 5; MEKK 5; Mekk5; mitogen activated protein kinase kinase kinase 5; mitogen-activated protein kinase kinase kinase 5; RGD1306565; RP3-325F22.4 | |
| Recombinant human MAP3K5 (immunogen range is 1191-1300). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| MAP3K5 |
| K52022_9A7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q99683 | |
| 4217 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title