missing translation for 'onlineSavingsMsg'
Learn More

ASK1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18425825 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425825 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425825 Supplier Novus Biologicals Supplier No. NBP176674AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASK1 Polyclonal antibody specifically detects ASK1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASK1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias Apoptosis signal-regulating kinase 1, ASK-1, ASK1MEKK 5, EC 2.7.11, MAP/ERK kinase kinase 5, MAPK/ERK kinase kinase 5, MAPKKK5EC 2.7.11.25, MEK kinase 5, MEKK5apoptosis signal regulating kinase 1, mitogen-activated protein kinase kinase kinase 5
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ASK1. The immunogen is located within amino acids 1260 - 1310 of ASK1. Amino Acid Squence: IEEKDQEIKHLKLKSQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4217
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.