Learn More
ASIC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA556148
Description
Immunogen sequence: EVQYVEKMEF PAVTFCNLNR FQTDAVAKFG VIFFLWHIVS KVLHLQEITA NSTGSREATD FAASHQNFSI VEFIRNKGFY LNNSTLLDCE FFGKPCSPKD FAHVFTEYGN CFTFNHGETL QAKRKVSVSG RGLSLLFNVN QEAFTDNPAL Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.
Cation channel that gives rise to very low constitutive currents in the absence of activation. The activated channel exhibits selectivity for sodium, and is inhibited by amiloride.
Specifications
| ASIC5 | |
| Polyclonal | |
| Unconjugated | |
| ASIC5 | |
| Acid-sensing ion channel 5, ASIC5, Amiloride-sensitive cation channel 5, Human intestine Na(+) channel, HINaC, ASIC5, ACCN5, HINAC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51802 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NY37 | |
| ASIC5 | |
| Recombit protein corresponding to Human ASIC5 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only