missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASIC3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577734
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
ACCN3 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, two hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. ACCN3 encodes a member that is an acid sensor and may play an important role in the detection of lasting pH changes. In addition, a heteromeric association between this member and ACCN1 has been observed as proton-gated channels sensitive to gadolinium.
Specifications
| ASIC3 | |
| Polyclonal | |
| Unconjugated | |
| ASIC3 | |
| ACCN3, DRASIC, Acid-sensing ion channel 3, amiloride-sensitive cation channel 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171209, 286920, 9311 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| O35240, Q6X1Y6, Q9UHC3 | |
| ASIC3 | |
| Peptide KPRSGLEEAQRRQAS(C), corresponding to amino acid residues 2-16 of rat ASIC3 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction