missing translation for 'onlineSavingsMsg'
Learn More

ASIC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15710352 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15710352 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15710352 Supplier Thermo Scientific Supplier No. PA562554

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ASIC2 Polyclonal Antibody

ASIC2 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ASIC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASIC2
Gene Accession No. Q16515
Gene Alias ACCN; ACCN1; ACIC2; acid sensing (proton gated) ion channel 2; acid sensing ion channel 2; acid sensing ion channel subunit 2; acid-sensing (proton-gated) ion channel 2; acid-sensing ion channel 2; Amiloride-sensitive brain sodium channel; amiloride-sensitive brain sodium channel 2; Amiloride-sensitive cation channel 1, neuronal; amiloride-sensitive cation channel 1, neuronal (degenerin); Amiloride-sensitive cation channel neuronal 1; Asic2; ASIC2a; Bnac1; BNaC1a; BNC1; BNC1k; Brain sodium channel 1; degenerin; hBNaC1; Mammalian degenerin homolog; MDEG; MDEG1; MDEG2; neuronal amiloride-sensitive cation channel 1
Gene Symbols ASIC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASIC2. Recombinant protein control fragment (Product #RP-101182).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 40
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.