missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ASIC1 Polyclonal Antibody

Brand: Thermo Scientific™ PA5109287
This item is not returnable.
View return policy
Description
Specifications
| ASIC1 | |
| Polyclonal | |
| Unconjugated | |
| ASIC1 | |
| Accn2; ACCN2 variant 3; acid sensing (proton gated) ion channel 1; acid sensing ion channel; acid sensing ion channel 1; acid sensing ion channel subunit 1; acid-sensing (proton-gated) ion channel 1; Acid-sensing ion channel; Acid-sensing ion channel 1; acid-sensing ion channel 1a protein; AI843610; Amiloride-sensitive cation channel 2, neuronal; Asic; ASIC1; ASIC1a; B530003N02Rik; BNAC2; brain sodium channel 2; Cation channel, amiloride-sensitive, neuronal, 2; degenerin 2; hBNaC2; proton gated cation channel ASIC1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 41 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P78348 | |
| ASIC1 | |
| Synthetic peptide within Human ACCN2 aa 457-506. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction