missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASIC1 Monoclonal Antibody (N271/44), FITC
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545593
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45593 was sufficient for detection of ASIC1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 60kDa. This antibody was formerly sold as clone S271-44.
This gene encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, 2 hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. The member encoded by this gene is expressed in most if not all brain neurons, and it may be an ion channel subunit; however, its function as an ion channel remains unknown. Cation channel with high affinity for sodium, which is gated by extracellular protons and inhibited by the diuretic amiloride. ASIC1 is permeable for Ca(2+), Li(+) and K(+) and is expressed in dorsal root ganglia, sciatic nerves, olfactory bulb, neo and allocortical regions, dentate granule cells, pyramidal cells of CA1-CA3 subfields of the hippocampal formation, habenula, basolateral amygdaloid nuclei, and in the Purkinje and granule cells of the cerebellum. It is diffusely detected over most other regions of the basal ganglia, including thalamic nuclei, substantia nigra, striatum and globus pallidus, hypothalamus, midbrain, pons, medulla and choroid plexus. Isoform 3 is expressed only in dorsal root ganglion while isoform 1 is expressed in DRG, spinal chord, trigeminal ganglia and the trigeminal mesencephalic nucleus.
Specifications
| ASIC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 | |
| P55926, P78348, Q6NXK8 | |
| ASIC1 | |
| Fusion protein amino acids 460-526 (Cytoplasmic C-terminus) of mouse ASIC1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N271/44 | |
| FITC | |
| ASIC1 | |
| Accn2; ACCN2 variant 3; acid sensing (proton gated) ion channel 1; acid sensing ion channel; acid sensing ion channel 1; acid sensing ion channel subunit 1; acid-sensing (proton-gated) ion channel 1; Acid-sensing ion channel; Acid-sensing ion channel 1; acid-sensing ion channel 1a protein; AI843610; Amiloride-sensitive cation channel 2, neuronal; Asic; ASIC1; ASIC1a; B530003N02Rik; BNAC2; brain sodium channel 2; Cation channel, amiloride-sensitive, neuronal, 2; degenerin 2; hBNaC2; proton gated cation channel ASIC1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 11419, 41, 79123 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction