missing translation for 'onlineSavingsMsg'
Learn More

Asiatic acid, 97%

Product Code. 15496618
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496618 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15496618 Supplier Thermo Scientific Alfa Aesar Supplier No. H60149.ME
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 464-92-6 | C30H48O5 | 488.709 g/mol

Asiatic acid is commonly used in wound healing. Asiatic acid has antioxidant, anti-inflammatory and neuroprotective properties. It is a starting material for asiatic acid derivative synthesis for use as anticancer agents, glycogen phosphorylase inhibitors and and hepatoprotectants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Asiatic acid is commonly used in wound healing. Asiatic acid has antioxidant, anti-inflammatory and neuroprotective properties. It is a starting material for asiatic acid derivative synthesis for use as anticancer agents, glycogen p
TRUSTED_SUSTAINABILITY

Specifications

CAS 464-92-6
Melting Point 325°C to 330°C
Molecular Formula C30H48O5
MDL Number MFCD00238541
Quantity 500 mg
Synonym Dammarolic acid
Solubility Information Soluble in dimethyl sulfoxide,ethanol and dimethylformamide. Sparingly soluble in water
InChI Key JXSVIVRDWWRQRT-YLPWNERUSA-N
SMILES CC1CCC2(CCC3(C(=CCC4C3(CCC5C4(CC(C(C5(C)CO)O)O)C)C)C2C1C)C)C(=O)O
IUPAC Name (4aS,6aS,6aS,6bR,8aS,9R,12aR,14bR)-10,11-dihydroxy-9-(hydroxymethyl)-1,2,6a,6b,9,12a-hexamethyl-2,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydro-1H-picene-4a-carboxylic acid
Molecular Weight (g/mol) 488.709
PubChem CID 129316838
Formula Weight 488.7
Percent Purity 97%
Chemical Name or Material Asiatic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.