missing translation for 'onlineSavingsMsg'
Learn More

Asialoglycoprotein Receptor 2 Antibody (OTI2A12), DyLight 594, Novus Biologicals™

Product Code. 18420815 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420815 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420815 Supplier Novus Biologicals Supplier No. NBP270212DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Asialoglycoprotein Receptor 2 Monoclonal antibody specifically detects Asialoglycoprotein Receptor 2 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Asialoglycoprotein Receptor 2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2A12
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias ASGPR 2, ASGP-R 2, ASGPR2, ASGP-R2, asialoglycoprotein receptor 2, CLEC4H2FLJ60040, C-type lectin domain family 4 member H2, HBxAg-binding protein, HBXBP, Hepatic lectin H2, HL-2
Host Species Mouse
Immunogen Full length recombinant protein of human Asialoglycoprotein Receptor 2 (NP_001172) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 433
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.