missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asialoglycoprotein Receptor 2 Antibody (002), Biotin , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-06211B
This item is not returnable.
View return policy
Description
Asialoglycoprotein Receptor 2 Monoclonal antibody specifically detects Asialoglycoprotein Receptor 2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Asialoglycoprotein Receptor 2 | |
| Monoclonal | |
| Biotin | |
| ASGPR 2, ASGP-R 2, ASGPR2, ASGP-R2, asialoglycoprotein receptor 2, CLEC4H2FLJ60040, C-type lectin domain family 4 member H2, HBxAg-binding protein, HBXBP, Hepatic lectin H2, HL-2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Asialoglycoprotein Receptor 2 (P07307-1; Gln80-Ala311). | |
| 0.1 mL | |
| Signal Transduction | |
| 433 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 002 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction