missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASHG/Fetuin A Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHAHSG

Code : NEW

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 15434893

  • £544.00 / 1 set
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

The Human ASHG/ Fetuin A ELISA quantitates Hu ASHG in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ASHG. Principle of the method The Human ASHG solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.
Specifications

Specifications

P02765
0.2 ng/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
197
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Inflammation
2°C to 8°C
Human
4 hr. 45 min.
0.205-50 ng/mL
0.2ng/mL to 50ng/mL
Biotin
Kinase & Inhibitors
This ELISA antibody pair detects human Fetuin A. Other species not determined.
Human
Colorimetric Microplate Reader
A2HS,AHS,FETUA,HSGA
<10%
HRP
RUO
Plasma, 1 μL; Serum, 1 μL; Supernatant, 100 μL
PRO2743, A2HS, AHS, FETUA, HSGA
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers