missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ASH2L Polyclonal antibody specifically detects ASH2L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications
Specifications
| Antigen | ASH2L |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | ASH2, ash2 (absent, small, or homeotic)-like (Drosophila), ASH2L1ash2 (absent, small, or homeotic, Drosophila, homolog)-like, ASH2L2ASH2-like protein, Bre2, set1/Ash2 histone methyltransferase complex subunit ASH2 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: TTGTTKKARSDPLFSAQRLPPHGYPLEHPFNKDGYRYILAEPDPHAPDPEKLELDCWAGKPIPGDLYRACLYERVLLALHDRAPQLKISDDRLTVVGEKGYSMVRASHGVRKGAWYFEITVDEM |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?