missing translation for 'onlineSavingsMsg'
Learn More

ASH2L Antibody, DyLight 488, Novus Biologicals™

Product Code. 18423833 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423833 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423833 Supplier Novus Biologicals Supplier No. NB600281G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASH2L Polyclonal antibody specifically detects ASH2L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASH2L
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ASH2, ash2 (absent, small, or homeotic)-like (Drosophila), ASH2L1ash2 (absent, small, or homeotic, Drosophila, homolog)-like, ASH2L2ASH2-like protein, Bre2, set1/Ash2 histone methyltransferase complex subunit ASH2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 575 and the C-terminus (residue 628) of human Absent, Small, or Homeotic-Like 2 using the numbering given in Swiss-Prot entry Q9UBL3 (GeneID 9070).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 9070
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.