missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASH2L Antibody (6F6B9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61685-0.025ml
This item is not returnable.
View return policy
Description
ASH2L Monoclonal specifically detects ASH2L in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ASH2L | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| ASH2, ash2 (absent, small, or homeotic)-like (Drosophila), ASH2L1ash2 (absent, small, or homeotic, Drosophila, homolog)-like, ASH2L2ASH2-like protein, Bre2, set1/Ash2 histone methyltransferase complex subunit ASH2 | |
| Mouse | |
| 71 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 6F6B9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ASH2L | |
| Purified recombinant fragment of human ASH2L (AA: 493-628) expressed in E. Coli. | |
| Protein G purified | |
| DNA Repair, Transcription Factors and Regulators | |
| 9070 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction