Learn More
ASGR2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11501-2-AP-20UL
Description
The ASGR2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ASGR2. This antibody recognizes human antigen. The ASGR2 antibody has been validated for the following applications: FC, IHC, IP, WB, ELISA analysis.
Specifications
| ASGR2 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 433 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Flow Cytometry, ELISA | |
| 227 μg/mL | |
| Rabbit | |
| Primary | |
| Human |