missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASGR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553410
This item is not returnable.
View return policy
Description
Immunogen sequence: YRHNYKNWAV TQPDNWHGHE LGGSEDCVEV QPDGRWNDDF CLQVYRWVCE KR Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 69%.
ASGR2 is a cell surface receptor that binds to galactose-terminated glycoproteins. It transports these glycoproteins via a series of membrane vesicles and tubules to an acidic-sorting organelle where the receptor and ligand dissociates. Then the receptor is recycled back to the cell surface. There are four alternatively spliced transcript variants of this gene. This gene has multiple polyadenylation sites.
Specifications
| ASGR2 | |
| Polyclonal | |
| Unconjugated | |
| ASGR2 | |
| ANCO1; ANCO-2; ASGPR 2; ASGP-R 2; ASGPR2; ASGP-R2; Asgr; Asgr2; Asgr-2; Asialoglycoprotein receptor 2; CLEC4H2; C-type lectin domain family 4 member H2; GAC-1; HBxAg-binding protein; HBXBP; Hepatic lectin 2; hepatic lectin 2 RHL2; hepatic lectin 2, RHL2; hepatic lectin H2; Hepatic lectin R2/3; HL-2; mHL-2; Nbla00144; RHL-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 433 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07307 | |
| ASGR2 | |
| Recombinant protein corresponding to Human ASGR2. Recombinant protein control fragment (Product #RP-90511). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction