missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASGR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568618
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 85%; Guinea Pig: 92%; Horse: 86%; Human: 100%; Mouse: 91%; Pig: 93%; Rat: 92%; Yeast: 75%.
The asialoglycooprotein receptor (ASGPR) is a transmembrane glycoprotein (42 kDa) which mediates binding, internalization and degradation of extracellular glycoproteins that have exposed terminal galactose residues. The receptor is expressed on the surface of the hepatocytes in a polar manner, i. e. it is present on the sinusoidal and lateral plasma membranes but not on the bile canalicular membrane. The mammalian hepatic ASGPR mediate the endocytosis and degradation of serum proteins from which terminal sialic residues have been removed. The rat asialoglycoprotein receptor (ASGPR) consists of three polypeptide subunits: rat hepatic lectin (RHL)-1, RHL-2 and RHL-3. The monoclonal antibody 8D7 recognizes a subunit-specific epitope of RHL-1 of rat ASGPR. The monoclonal antibody 8D7 is cross reactive with human ASGPR.
Specifications
| ASGR1 | |
| Polyclonal | |
| Unconjugated | |
| Asgr1 | |
| ASGPR; ASGPR 1; ASGP-R 1; ASGPR1; ASGR; ASGR1; Asgr-1; asialoglycoprotein receptor (RHL1); asialoglycoprotein receptor 1; Asialoglycoprotein receptor 1 (hepatic lectin); asialoglycoprotein receptor RHL1; CLEC4H1; C-type lectin domain family 4 member H1; Hepatic lectin 1; hepatic lectin 1 RHL1; hepatic lectin 1, RHL1; hepatic lectin H1; HL-1; RATRHL1; RHL1; RHL-1; Unknown (protein for MGC:133691) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 432 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P07306 | |
| Asgr1 | |
| synthetic peptide directed towards the middle region of human ASGR1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction