missing translation for 'onlineSavingsMsg'
Learn More

ASGPR1 Antibody (024), HRP, Novus Biologicals™

Product Code. 18343230 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343230 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343230 Supplier Novus Biologicals Supplier No. NBP289638H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ASGPR1 Monoclonal specifically detects ASGPR1 in Human samples. It is validated for Western Blot, Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASGPR1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 24
Conjugate HRP
Formulation PBS
Gene Alias ASGPR, ASGPR 1, ASGP-R 1, asialoglycoprotein receptor 1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ASGPR1 (P07306; Gln62-Leu291).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 432
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.