missing translation for 'onlineSavingsMsg'
Learn More

ASF1b Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18341433 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341433 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341433 Supplier Novus Biologicals Supplier No. NBP300286AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASF1b Polyclonal specifically detects ASF1b in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASF1b
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ASF1 silencing function 1 homolog B (S. cerevisiae), CCG1-interacting factor A-II, CIA-II, FLJ10604, hAsf1, hAsf1b, hCIA-II, histone chaperone ASF1B, silencing function protein 1 homolog B
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ASF1b (Q9NVP2; Met1-Ile202).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 55723
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.