missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASF1b Antibody (128), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90280CL1
This item is not returnable.
View return policy
Description
ASF1b Monoclonal antibody specifically detects ASF1b in Human samples. It is validated for ELISA
Specifications
| ASF1b | |
| Monoclonal | |
| CoraFluor 1 | |
| ASF1 silencing function 1 homolog B (S. cerevisiae), CCG1-interacting factor A-II, CIA-II, FLJ10604, hAsf1, hAsf1b, hCIA-II, histone chaperone ASF1B, silencing function protein 1 homolog B | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ASF1b (Uniprot#: Q9NVP2; Met1-Ile202). | |
| 0.1 mL | |
| Cancer, DNA replication Transcription Translation and Splicing | |
| 55723 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 128 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction