missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASF1a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | ASF1a |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18201332
|
Novus Biologicals
NBP2-57608 |
100 μL |
£435.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685636
|
Novus Biologicals
NBP2-57608-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASF1a Polyclonal specifically detects ASF1a in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| ASF1a | |
| Polyclonal | |
| Rabbit | |
| DNA Repair | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 25842 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DSVLVGPVPAGRHMFVFQADAPNPGLIPDADAVGVTVVLITCTYRGQEFIRVGYYVNNEYTETELRENPPVKPDFSKLQRNILASNPRVTRFHINWEDNTEKLEDAESSNPNLQSLLSTDALPSASKGWSTSENSLNVMLESHM | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| ASF1 silencing function 1 homolog A (S. cerevisiae), CCG1-interacting factor A, CIACGI-98, DKFZP547E2110, hAsf1, hAsf1a, hCIA, histone chaperone ASF1A, HSPC146, silencing function 1A, silencing function protein 1 homolog A | |
| ASF1A | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title