Learn More
CPC (Colder) AseptiQuik™ STC II Genderless Quick-Disconnect Fittings, AspetiQuik™ S to Sanitary Adapter, Polycarbonate
Product Code. 30053916
Click to view available options
Diameter (English) Inner:
1-1/2 in.
3/4 in.
Unit Size:
Each
Product Code. 30053916
Brand: CPC (Colder) 5011204
Only null left
Add to basket
Add to basket
Low-flow genderless sterile connector and Steam-Thru™ II connector in a single unit simplify process integration while maximizing flexibility
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. Simple flip, click, and pull design minimizes operator errors. Create quick and easy sterile connections, even in nonsterile environments. Robust construction provides reliable and repeatable fluid transfer. Specifically designed for small-volume fluid transfer applications. Innovative three-port steam design. Sterilize AseptikQuik STC II fitting on stainless steel equipment via SIP. 0.375 in. nominal flow path size
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1-1/2 in.
Min. Operating Temperature (Metric) 1°C
Packaging Quantity 1 Ea.
Outlet Port Quick Disconnect x Sanitary x Sanitary
Sterilization Method 130°C
Shape Straight
Includes 3/4 in. Sanitary Clamp Size
Material (Seal) Platinum-cured Silicone
Color White with blue pull tab and amber tint