Learn More
CPC (Colder) AseptiQuik™ S Low-Flow Genderless Quick-Disconnect Fittings, Hose Barb Adapter, Polycarbonate
Product Code. 30053496 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/4 in.
1/8 in.
3/8 in.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner unitSize
30053496 1/8 in. Each
30053471 3/8 in. Each
30052995 1/4 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30053496 Supplier CPC (Colder) Supplier No. AQS17002
Only null left
Add to basket
Add to basket
Low-flow genderless sterile connectors simplify process integration while maximizing flexibility
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. Simple flip, click, and pull design minimizes operator errors. Create quick and easy sterile connections, even in nonsterile environments. Specifically designed for small-volume fluid transfer applications. Robust construction provides reliable and repeatable fluid transfer. 0.25 in. nominal flow path size
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1/8 in.
Min. Operating Temperature (Metric) 1°C
Packaging Quantity 1 Ea.
Outlet Port Quick Disconnect x Hose Barb
Sterilization Method Gamma irradiation up to 50 kGy
Shape Straight
Material (Seal) Platinum-cured Silicone
Color White with blue pull tab