missing translation for 'onlineSavingsMsg'
Learn More

Asenapine N-Oxide, TRC

Product Code. 30134238
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134238 2.5 mg 2.5mg
30134701 10 mg 10mg
30006373 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134238 Supplier Toronto Research Chemicals Supplier No. TRCA7880102.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128949-51-9
Molecular Formula C17 H16 Cl N O2
Molecular Weight (g/mol) 301.77
Synonym (3aR-trans)-5-Chloro-2,3,3a,12b-tetrahydro-2-methyl-1H-dibenz[2,3:6,7]oxepino[4,5-c]pyrrole 2-Oxide;
IUPAC Name (2R,6R)-9-chloro-4-methyl-4-oxido-13-oxa-4-azoniatetracyclo[12.4.0.02,6.07,12]octadeca-1(18),7(12),8,10,14,16-hexaene
SMILES C[N+]1([O-])CC2C(C1)c3cc(Cl)ccc3Oc4ccccc24

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C17H16ClNO2/c1-19(20)9-14-12-4-2-3-5-16(12)21-17-7-6-11(18)8-13(17)15(14)10-19/h2-8,14-15H,9-10H2,1H3
Formula Weight 301.087
Chemical Name or Material Asenapine N-oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.