missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asenapine, MedChemExpress
Description
Asenapine (Org 5222), an atypical antipsychotic, is an antagonist of serotonin receptors (pKi: 8.4-10.5), adrenoceptors (pKi: 8.9-9.5), dopamine receptors (pKi: 8.9-9.4) and histamine receptors (pKi: 8.2-9.0). Asenapine can be used in the research of schizophrenia and bipolar disorder.
Specifications
Specifications
| Chemical Name or Material | Asenapine |
| CAS | 65576-45-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H16ClNO |
| Quantity | 100 mg |
| Synonym | Org 5222 |
| Solubility Information | DMSO : 200 mg/mL (699.86 mM; Need ultrasonic) |
| SMILES | ClC1=CC=C2OC3=CC=CC=C3[C@@](C4)([H])[C@](CN4C)([H])C2=C1 |
| Molecular Weight (g/mol) | 285.77 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?