missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asenapine Maleate, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Asenapine maleate |
| CAS | 85650-56-2 |
| Molecular Formula | C17 H16 Cl N O . C4 H4 O4 |
| Quantity | 10 mg |
| Synonym | 1H-Dibenz[2,3:6,7]oxepino[4,5-c]pyrrole, 5-chloro-2,3,3a,12b-tetrahydro-2-methyl-, (3aR,12bR)-rel-, (2Z)-2-butenedioate (1:1), 1H-Dibenz[2,3:6,7]oxepino[4,5-c]pyrrole, 5-chloro-2,3,3a,12b-tetrahydro-2-methyl-, trans-, (Z)-2-butenedioate (1:1), Asenapine maleate, Org 5222, rel-(3aR,12bR)-5-Chloro-2,3,3a,12b-tetrahydro-2-methyl-1H-dibenz[2,3:6,7]oxepino[4,5-c]pyrrole maleate |
| InChI Formula | InChI=1S/C17H16ClNO.C4H4O4/c1-19-9-14-12-4-2-3-5-16(12)20-17-7-6-11(18)8-13(17)15(14)10-19;5-3(6)1-2-4(7)8/h2-8,14-15H,9-10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-,15-;/m1./s1 |
| SMILES | CN1C[C@H]2[C@H](C1)c3cc(Cl)ccc3Oc4ccccc24.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;(2S,6S)-9-chloro-4-methyl-13-oxa-4-azatetracyclo[12.4.0.02,6.07,12]octadeca-1(18),7(12),8,10,14,16-hexaene |
| Molecular Weight (g/mol) | 401.84 |
| Formula Weight | 401.103 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction