missing translation for 'onlineSavingsMsg'
Learn More

Asenapine maleate, MedChemExpress

Product Code. 16451596
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16451596

Brand: Medchem Express HY11100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Asenapine maleate is a 5-HT (1A, 1B, 2A, 2B, 2C, 5A, 6, 7) and D2 antagonist with Ki values of 0.03-4.0 nM, 1.3nM, respectively, and an antipsychotic.

Asenapine maleate is a 5-HT (1A, 1B, 2A, 2B, 2C, 5A, 6, 7) and D2 antagonist with Ki values of 0.03-4.0 nM, 1.3nM, respectively, and an antipsychotic.

Specifications

Chemical Name or Material Asenapine maleate
CAS 85650-56-2
Color White
Purity Grade Notes Research
Molecular Formula C21H20ClNO5
Quantity 500 mg
Synonym Org 5222 maleate
Solubility Information DMSO : 25 mg/mL (62.21 mM; Need ultrasonic) ∣H2O : 6.25 mg/mL (15.55 mM; Need ultrasonic and warming)
SMILES CN1C[C@]2([H])C3=CC=CC=C3OC4=CC=C(Cl)C=C4[C@]2(C1)[H].O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 401.84
Formula Weight 401.84
Percent Purity 95.07%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.