missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Asenapine maleate
GREENER_CHOICE

Product Code. 18702544
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18702544

Brand: Tocris Bioscience™ 3737/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-selective 5-HT, dopamine, adrenalin and histamine receptor antagonist; antipsychotic agent

Asenapine maleate is a novel psychopharmacologic agent. Displays antagonist activity at 5-HT, dopamine, noradrenalin and histamine receptor subtypes (pKi values are 8.60, 8.40, 10.15, 9.75, 10.46, 8.84, 9.60, 9.94, 8.85, 8.90, 8.84, 9.38, 8.95, 8.93, 8.9, 9.49, 8.91, 9.00 and 8.21 for 5-HT1A, 5-HT1B, 5-HT2A, 5-HT2B, 5-HT2C, 5-HT5A, 5-HT6, 5-HT7, D1, D2L, D2S, D3, D4, α1A, α2A, α2B, α2C, H1 and H2 receptors respectively). Displays no appreciable affinity for muscarinic receptors. Exhibits potent activity in animal models predictive of antipsychotic efficacy.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type Asenapine maleate
Purity >98%
Molecular Weight (g/mol) 401.84
Inhibitors Asenapine maleate

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.